Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103281.2 | NP_001096751.1 | 1 - 2 | 50 | 66 | |
GenBank mRNA | BC149711.1 | - | 1 - 2 | 51 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |