Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP11L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105431.1 | NP_001098901.1 | 1 - 2 | 106 | 52 | |
GenBank mRNA | BC153231.1 | - | 1 - 2 | 106 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |