Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105455.1 | NP_001098925.1 | 1 | 157 | 117 | |
GenBank mRNA | BC102562.1 | - | 1 | 157 | 117 | |
BC151476.1 | - | 1 | 154 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |