Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110803.2 | NP_001104273.2 | 7 - 8 | 1433 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |