Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113314.1 | NP_001106785.1 | 4 - 5 | 242 | 82 | |
GenBank mRNA | BC151397.1 | - | 4 - 5 | 242 | 82 | |
M99393.1 | - | 1 - 2 | 127 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |