Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098157.1 | NP_001091626.1 | 1 - 2 | 129 | 94 | |
GenBank mRNA | BC140609.1 | - | 1 - 2 | 129 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |