Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099210.2 | NP_001092680.1 | 4 - 5 | 365 | 96 | |
GenBank mRNA | BC142374.1 | - | 4 - 5 | 367 | 96 | |
BC148018.1 | - | 4 - 5 | 349 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |