Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101294.2 | NP_001094764.2 | 2 - 3 | 1129 | 55 | |
GenBank mRNA | BC148065.1 | - | 2 - 3 | 1129 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |