Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105050.1 | NP_001098520.1 | 3 - 4 | 245 | 70 | |
GenBank mRNA | BC126742.1 | - | 3 - 4 | 245 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |