Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105472.1 | NP_001098942.1 | 89 | 70 | ||
GenBank mRNA | BC149174.1 | - | 89 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus |