Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111262.2 | NP_001104732.1 | 4 - 5 | 876 | 150 | |
GenBank mRNA | EF446903.1 | - | 4 - 5 | 877 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |