Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111262.2 | NP_001104732.1 | 5 - 6 | 901 | 102 | |
GenBank mRNA | EF446903.1 | - | 5 - 6 | 902 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |