Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111263.1 | NP_001104733.1 | 1 - 2 | 45 | 74 | |
GenBank mRNA | EF446906.1 | - | 1 - 2 | 45 | 74 | |
EF446907.1 | - | 1 - 2 | 45 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |