Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109978.1 | NP_001103448.1 | 2 - 3 | 220 | 157 | |
NM_176618.2 | NP_788791.1 | 2 - 3 | 250 | 157 | ||
GenBank mRNA | AB099026.1 | - | 1 - 2 | 58 | 157 | |
AF020509.1 | - | 2 - 3 | 250 | 157 | ||
BC133469.1 | - | 2 - 3 | 258 | 157 | ||
EU107962.1 | - | 2 - 3 | 220 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |