Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176626.1 | NP_788799.1 | 3 - 4 | 318 | 123 | |
GenBank mRNA | AF192335.1 | - | 3 - 4 | 318 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |