Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176628.1 | NP_788801.1 | 8 - 9 | 994 | 141 | |
GenBank mRNA | AF192336.1 | - | 8 - 9 | 994 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Bos taurus Probe spans exons |