Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174411.2 | NP_776836.1 | 7 - 8 | 912 | 159 | |
GenBank mRNA | AB098781.2 | - | 2 - 3 | 405 | 159 | |
BC134743.1 | - | 7 - 8 | 941 | 159 | ||
M73962.1 | - | 7 - 8 | 912 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |