Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014938.1 | NP_001014938.1 | 3 | 2256 | 140 | |
GenBank mRNA | BT021877.1 | - | 3 | 2256 | 140 | |
DQ164537.1 | - | 3 | 2257 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |