Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105014.2 | NP_001098484.1 | 5 | 1445 | 116 | |
GenBank mRNA | BC150032.1 | - | 5 | 1446 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |