Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174493.1 | NP_776918.1 | 2 | 1044 | 149 | |
GenBank mRNA | BC120043.1 | - | 2 | 1045 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |