Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMKLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145235.1 | NP_001138707.1 | 1 | 950 | 80 | |
GenBank mRNA | FJ594407.1 | - | 1 | 950 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |