Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100371.1 | NP_001093841.1 | 5 - 6 | 1856 | 77 | |
GenBank mRNA | BC149232.1 | - | 5 - 6 | 1856 | 77 | |
BT026149.1 | - | 1 - 2 | 660 | 77 | ||
BT026332.1 | - | 1 - 2 | 620 | 77 | ||
BT029787.1 | - | 1 - 2 | 665 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |