Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166515.1 | NP_001159987.1 | 1 - 2 | 1116 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |