Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTNL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103345.1 | NP_001096815.1 | 9 - 10 | 1140 | 113 | |
GenBank mRNA | BC149479.1 | - | 9 - 10 | 1140 | 113 | |
BT029803.1 | - | 9 - 10 | 1161 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |