Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105497.2 | NP_001098967.1 | 3 - 4 | 247 | 63 | |
GenBank mRNA | BC116154.1 | - | 3 - 4 | 248 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |