Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163458.1 | NP_001156930.1 | 6 - 7 | 1418 | 73 | |
GenBank mRNA | BC126681.1 | - | 6 - 7 | 1358 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |