Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163458.1 | NP_001156930.1 | 9 - 10 | 1855 | 67 | |
GenBank mRNA | BC126681.1 | - | 9 - 10 | 1795 | 67 | |
DV911405.1 | - | 5 - 6 | 461 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |