Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163785.1 | NP_001157257.1 | 3 - 4 | 859 | 83 | |
GenBank mRNA | EF690283.1 | - | 3 - 4 | 859 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |