Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163935.1 | NP_001157407.1 | 2 - 3 | 378 | 92 | |
GenBank mRNA | EE368568.1 | - | 1 - 2 | 73 | 92 | |
EH149935.1 | - | 3 - 4 | 496 | 92 | ||
EH165066.1 | - | 2 - 3 | 378 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |