Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205470.1 | NP_001192399.1 | 19 - 20 | 2035 | 83 | |
GenBank mRNA | BC151667.1 | - | 19 - 20 | 2007 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |