Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205478.1 | NP_001192407.1 | 8 - 9 | 1604 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |