Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205546.3 | NP_001192475.1 | 5 - 6 | 805 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |