Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205751.2 | NP_001192680.2 | 2 - 3 | 1181 | 84 | |
GenBank mRNA | BC148920.1 | - | 2 - 3 | 913 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |