Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205971.1 | NP_001192900.1 | 1 - 2 | 564 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |