Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206053.2 | NP_001192982.1 | 13 - 14 | 2015 | 75 | |
GenBank mRNA | BC104621.1 | - | 8 - 9 | 1020 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |