Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206097.2 | NP_001193026.2 | 12 - 13 | 1378 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |