Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206124.2 | NP_001193053.1 | 5 - 6 | 1067 | 127 | |
NM_001354592.1 | NP_001341521.1 | 5 - 6 | 1041 | 127 | ||
GenBank mRNA | BC109916.1 | - | 5 - 6 | 929 | 127 | |
EH374978.1 | - | 4 - 5 | 656 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |