Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206217.1 | NP_001193146.1 | 2 - 3 | 1065 | 91 | |
GenBank mRNA | BT025396.1 | - | 2 - 3 | 1065 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |