Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206217.1 | NP_001193146.1 | 6 - 7 | 2405 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |