Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206238.1 | NP_001193167.1 | 2 - 3 | 840 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Probe spans exons |