Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYNRIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192459.2 | NP_001179388.1 | 3 - 4 | 993 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |