Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYNRIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192459.2 | NP_001179388.1 | 4 - 5 | 2516 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |