Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192494.1 | NP_001179423.1 | 9 - 10 | 1215 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |