Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H5orf46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206334.1 | NP_001193263.1 | 2 - 3 | 254 | 110 | |
GenBank mRNA | BC102132.1 | - | 2 - 3 | 256 | 110 | |
EV681338.1 | - | 2 - 3 | 258 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |