Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206440.1 | NP_001193369.1 | 1 - 2 | 209 | 83 | |
GenBank mRNA | BC151821.1 | - | 1 - 2 | 210 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |