Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206471.1 | NP_001193400.1 | 2824 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus |