Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192672.3 | NP_001179601.1 | 2 - 3 | 416 | 98 | |
GenBank mRNA | AB508930.1 | - | 1 - 2 | 237 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |