Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192781.2 | NP_001179710.1 | 4 - 5 | 555 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |