Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163946.2 | NP_001157418.1 | 6 - 7 | 683 | 74 | |
GenBank mRNA | BC149694.1 | - | 6 - 7 | 684 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |