Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206676.1 | NP_001193605.1 | 2 - 3 | 355 | 76 | |
GenBank mRNA | AF185291.1 | - | 1 - 2 | 81 | 76 | |
GH309407.1 | - | 2 - 3 | 355 | 76 | ||
GH315554.1 | - | 1 - 2 | 68 | 76 | ||
LC223005.1 | - | 2 - 3 | 390 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |